Methyl sulfoxide-d6, for NMR, 100.0 atom % D, min. 99.96 atom % D
Methyl sulfoxide-d6, for NMR, 100.0 atom % D, min. 99.96 atom % D
Methyl sulfoxide-d6, for NMR, 100.0 atom % D, min. 99.96 atom % D
Methyl sulfoxide-d6, for NMR, 100.0 atom % D, min. 99.96 atom % D
Methyl sulfoxide-d6, for NMR, 100.0 atom % D, min. 99.96 atom % D
CAS: 2206-27-1 | C2H6OS | 84.17 g/mol
Have Questions?
Catalog number FSA169020250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS593-45-3
IUPAC Nameoctadecane
Molecular FormulaC18H38
InChI KeyRZJRJXONCZWCBN-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solid or liquid
Infrared spectrumConforms
Melting point28°C to 31°C
Appearance (Color)White to yellow
GC>=99.0 %
For NMR
RUO – Research Use Only