Potassium tert-butoxide, 98+%, pure, Thermo Scientific Chemicals
Potassium tert-butoxide, 98+%, pure, Thermo Scientific Chemicals
Potassium tert-butoxide, 98+%, pure, Thermo Scientific Chemicals
Potassium tert-butoxide, 98+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Potassium tert-butoxide, 98+%, pure, Thermo Scientific Chemicals
CAS: 865-47-4 | C4H9KO | 112.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Packaging:
View all
Glass Bottle
Catalog number FSA168880050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium tert-butoxide
CAS865-47-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Flammable solid.
Self-heating in large quantities; may catch fire.
Reacts violently with water.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several min