Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol, Thermo Scientific Chemicals
Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol, Thermo Scientific Chemicals
Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol, Thermo Scientific Chemicals
Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol, Thermo Scientific Chemicals
CAS: 124-41-4 | CH3NaO | 54.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
1 L
2.5 L
10 L
Catalog number FSA168600250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Request bulk or custom format
Chemical Identifiers
CAS30652-11-0
IUPAC Name3-hydroxy-1,2-dimethyl-1,4-dihydropyridin-4-one
Molecular FormulaC7H9NO2
InChI KeyTZXKOCQBRNJULO-UHFFFAOYSA-N
SMILESCN1C=CC(=O)C(O)=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or needles
Infrared spectrumConforms
TLC>=99.0 %
Heavy metals=<20 ppm
pH6 to 8 (1 % solution)