Hexanes, 95+%, for spectroscopy, mixture of isomers, Thermo Scientific Chemicals
Hexanes, 95+%, for spectroscopy, mixture of isomers, Thermo Scientific Chemicals
Hexanes, 95+%, for spectroscopy, mixture of isomers, Thermo Scientific Chemicals
Hexanes, 95+%, for spectroscopy, mixture of isomers, Thermo Scientific Chemicals
Hexanes, 95+%, for spectroscopy, mixture of isomers, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hexanes, 95+%, for spectroscopy, mixture of isomers, Thermo Scientific Chemicals
CAS: 92112-69-1 | C6H14 | 86.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Packaging:
Glass bottle
Catalog number FSA168570025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialHexanes
Acidity0.00015 meq/g max.
CAS92112-69-1
Absorbance(1 cm cell vs water), 0.005 max. at 240nm, 0.005 max. at 250nm, 0.01 max. at 230nm, 0.025 max. at 220nm, 0.1 max. at 210nm, 0.3 max. at 200nm
Health Hazard 1GHS Signal Word: Danger
View more
For UV/VIS spectroscopy
RUO – Research Use Only