View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialIsoamyl nitrite
Name Note97%
CAS110-46-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement: Harmful if inhaled. Harmful if swallowed. Highly flammable liquid and vapor.