N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, 99+%, Thermo Scientific Chemicals
N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, 99+%, Thermo Scientific Chemicals
N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, 99+%, Thermo Scientific Chemicals
N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, 99+%, Thermo Scientific Chemicals
CAS: 16357-59-8 | C14H17NO3 | 247.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Catalog number FSA165351000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS149-74-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.5180 to 1.5200 (20°C, 589 nm)
Appearance (Form)Liquid
Coupling reagent
RUO – Research Use Only