5-Methylsalicylic acid, 98%
5-Methylsalicylic acid, 98%
5-Methylsalicylic acid, 98%
5-Methylsalicylic acid, 98%
Thermo Scientific Chemicals
5-Methylsalicylic acid, 98%
CAS: 89-56-5 | C8H8O3 | 152.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA16386010010 gGlass bottle
Catalog number FSA163860100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS609-99-4
IUPAC Name2-hydroxy-3,5-dinitrobenzoic acid
Molecular FormulaC7H4N2O7
InChI KeyLWFUFLREGJMOIZ-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(=CC(=C1O)[N+]([O-])=O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=97.5 %
Appearance (Color)Yellow
Appearance (Form)Powder
Infrared spectrumConforms
Melting point170°C to 174°C