1,3-Dicyanobenzene, 98%, Thermo Scientific Chemicals
1,3-Dicyanobenzene, 98%, Thermo Scientific Chemicals
1,3-Dicyanobenzene, 98%, Thermo Scientific Chemicals
1,3-Dicyanobenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,3-Dicyanobenzene, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Plastic bottle
Catalog number FSA163662500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS827-36-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.5110-1.5160 @ 20°C