Brucine dihydrate, 99%, Thermo Scientific Chemicals
Brucine dihydrate, 99%, Thermo Scientific Chemicals
Brucine dihydrate, 99%, Thermo Scientific Chemicals
Brucine dihydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Brucine dihydrate, 99%, Thermo Scientific Chemicals
CAS: 145428-94-0 | C23H27N2O4 | 395.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number FSA163500500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7440-06-4
IUPAC Nameplatinum
Molecular FormulaPt
InChI KeyBASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES[Pt]
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on Drying45-60% w/w
Assay from Suppliers CofAPt: 4.85-5.15% w/w