α-Bromo-p-tolunitrile, 98%, Thermo Scientific Chemicals
α-Bromo-p-tolunitrile, 98%, Thermo Scientific Chemicals
α-Bromo-p-tolunitrile, 98%, Thermo Scientific Chemicals
α-Bromo-p-tolunitrile, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
α-Bromo-p-tolunitrile, 98%, Thermo Scientific Chemicals
CAS: 17201-43-3 | C8H6BrN | 196.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA162760100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS867-55-0
IUPAC Namelithium(1+) 2-hydroxypropanoate
Molecular FormulaC3H5LiO3
InChI KeyGKQWYZBANWAFMQ-UHFFFAOYNA-M
SMILES[Li+].CC(O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration with HClO498.5 to 101.5 %
Appearance (Form)Crystalline powder
pH6 to 9 (1% solution)
Solubility(10% in water) Clear colorless