Phenyl benzoate, 99%, Thermo Scientific Chemicals
Phenyl benzoate, 99%, Thermo Scientific Chemicals
Phenyl benzoate, 99%, Thermo Scientific Chemicals
Phenyl benzoate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenyl benzoate, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA162061000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS18883-66-4
IUPAC Name3-methyl-3-nitroso-1-[(2S,3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]urea
Molecular FormulaC8H15N3O7
InChI KeyZSJLQEPLLKMAKR-GKHCUFPYSA-N
SMILESCN(N=O)C(=O)N[C@H]1[C@@H](O)O[C@H](CO)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)alpha-anomer: 75.0% min.
Appearance (Color)White to pale yellow
Assay (HPLC)>97.0%