2-Amino-5-methylbenzoic acid, 97%, Thermo Scientific Chemicals
2-Amino-5-methylbenzoic acid, 97%, Thermo Scientific Chemicals
2-Amino-5-methylbenzoic acid, 97%, Thermo Scientific Chemicals
2-Amino-5-methylbenzoic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Amino-5-methylbenzoic acid, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA160200050
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS19004-19-4
IUPAC Namedicopper(2+) pentahydrate tetranitrate
Molecular FormulaCu2H10N4O17
InChI KeyIZDJJEMZQZQQQQ-UHFFFAOYSA-N
SMILESO.O.O.O.O.[Cu++].[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride (Cl)=<20 ppm
Calcium (Ca)=<50 ppm
Lead (Pb)=<10 ppm
Potassium (K)=<50 ppm
Appearance (Color)Blue