2,4-Di-tert-butylphenol, 97%, Thermo Scientific Chemicals
2,4-Di-tert-butylphenol, 97%, Thermo Scientific Chemicals
2,4-Di-tert-butylphenol, 97%, Thermo Scientific Chemicals
2,4-Di-tert-butylphenol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,4-Di-tert-butylphenol, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Glass bottle
Catalog number FSA160125000
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7440-32-6
IUPAC Nametitanium
Molecular FormulaTi
InChI KeyRTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES[Ti]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.2% (metals basis)
Appearance (Color)Silvery.
FormFoil, 1.0mm (0.040in) thick.