Triphenylphosphine Sulfide, 99+%
Triphenylphosphine Sulfide, 99+%
Triphenylphosphine Sulfide, 99+%
Triphenylphosphine Sulfide, 99+%
Thermo Scientific Chemicals
Triphenylphosphine Sulfide, 99+%
CAS: 3878-45-3 | C18H15PS | 294.35 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA15903010010 gGlass bottle
Catalog number FSA159030100
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS624-92-0
IUPAC Name(methyldisulfanyl)methane
Molecular FormulaC2H6S2
InChI KeyWQOXQRCZOLPYPM-UHFFFAOYSA-N
SMILESCSSC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
Infrared spectrumConforms
Refractive index1.5243 to 1.5263 (20°C, 589 nm)
GC>=98.5 %
Water=<0.06 %