Boron trifluoride dimethanol complex, 50-52 wt% BF3, Thermo Scientific Chemicals
Boron trifluoride dimethanol complex, 50-52 wt% BF3, Thermo Scientific Chemicals
Boron trifluoride dimethanol complex, 50-52 wt% BF3, Thermo Scientific Chemicals
Boron trifluoride dimethanol complex, 50-52 wt% BF3, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Boron trifluoride dimethanol complex, 50-52 wt% BF3, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
50 g
250 g
Packaging:
Glass bottle
Catalog number FSA158900010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS109-04-6
IUPAC Name2-bromopyridine
Molecular FormulaC5H4BrN
InChI KeyIMRWILPUOVGIMU-UHFFFAOYSA-N
SMILESBrC1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow or pale brown
FormLiquid
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.5695-1.5725 @ 20?C
Lewis acid
RUO – Research Use Only