1-Methyl-4-piperidone, 98%, Thermo Scientific Chemicals
1-Methyl-4-piperidone, 98%, Thermo Scientific Chemicals
1-Methyl-4-piperidone, 98%, Thermo Scientific Chemicals
1-Methyl-4-piperidone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Methyl-4-piperidone, 98%, Thermo Scientific Chemicals
CAS: 1445-73-4 | C6H11NO | 113.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA156921000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS24425-40-9
IUPAC Name2,3-dihydro-1H-inden-5-amine
Molecular FormulaC9H11N
InChI KeyLEWZOBYWGWKNCK-UHFFFAOYSA-N
SMILESNC1=CC=C2CCCC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=96.0 %
Appearance (Color)Brown
Appearance (Form)Crystalline mass