1,4-Butanediol diglycidyl ether, 60%, technical, Thermo Scientific Chemicals
1,4-Butanediol diglycidyl ether, 60%, technical, Thermo Scientific Chemicals
1,4-Butanediol diglycidyl ether, 60%, technical, Thermo Scientific Chemicals
1,4-Butanediol diglycidyl ether, 60%, technical, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,4-Butanediol diglycidyl ether, 60%, technical, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
Catalog number FSA154310025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 kg
Request bulk or custom format
Chemical Identifiers
CAS4412-96-8
IUPAC Name3-methylfuran-2-carboxylic acid
Molecular FormulaC6H6O3
InChI KeyBNYIQEFWGSXIKQ-UHFFFAOYSA-N
SMILESCC1=C(OC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or granules
Appearance (Color)White to cream
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Assay (Silylated GC)≥98.0%