9-Anthracenecarboxaldehyde, 99%, Thermo Scientific Chemicals
9-Anthracenecarboxaldehyde, 99%, Thermo Scientific Chemicals
9-Anthracenecarboxaldehyde, 99%, Thermo Scientific Chemicals
9-Anthracenecarboxaldehyde, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
9-Anthracenecarboxaldehyde, 99%, Thermo Scientific Chemicals
CAS: 642-31-9 | C15H10O | 206.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA153931000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS102-49-8
IUPAC Name1-(3,4-dichlorophenyl)methanamine
Molecular FormulaC7H7Cl2N
InChI KeyIXHNFOOSLAWRBQ-UHFFFAOYSA-N
SMILESNCC1=CC=C(Cl)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
GC>=97.5 %
Infrared spectrumConforms
Refractive index1.5775 to 1.5795 (20°C, 589 nm)
Appearance (Form)Liquid