tert-Butylhydroquinone, 97%, Thermo Scientific Chemicals
tert-Butylhydroquinone, 97%, Thermo Scientific Chemicals
tert-Butylhydroquinone, 97%, Thermo Scientific Chemicals
tert-Butylhydroquinone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
tert-Butylhydroquinone, 97%, Thermo Scientific Chemicals
CAS: 1948-33-0 | C10H14O2 | 166.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
250 g
Packaging:
View all
Plastic bottle
Catalog number FSA150825000
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7782-42-5
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.2% max.