Sulfanilic acid, 99%, Thermo Scientific Chemicals
Sulfanilic acid, 99%, Thermo Scientific Chemicals
Sulfanilic acid, 99%, Thermo Scientific Chemicals
Sulfanilic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sulfanilic acid, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA150711000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS67-56-1
IUPAC Namemethanol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-UHFFFAOYSA-N
SMILESCO
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.8 %
Acidity/alkalinity=<0.0005 meq/g
Appearance (Form)Clear liquid
Infrared spectrumConforms
Water=<0.005 % (Coulometric)