L(-)-Malic acid, 99%, Thermo Scientific Chemicals
L(-)-Malic acid, 99%, Thermo Scientific Chemicals
L(-)-Malic acid, 99%, Thermo Scientific Chemicals
L(-)-Malic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L(-)-Malic acid, 99%, Thermo Scientific Chemicals
CAS: 97-67-6 | C4H6O5 | 134.087 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Plastic bottle
Catalog number FSA150595000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS79099-07-3
IUPAC Nametert-butyl 4-oxopiperidine-1-carboxylate
Molecular FormulaC10H17NO3
InChI KeyROUYFJUVMYHXFJ-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)N1CCC(=O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
Melting Point (clear melt)70.5-76.5?C