2,3-Butanedione monoxime, 98%, Thermo Scientific Chemicals
2,3-Butanedione monoxime, 98%, Thermo Scientific Chemicals
2,3-Butanedione monoxime, 98%, Thermo Scientific Chemicals
2,3-Butanedione monoxime, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,3-Butanedione monoxime, 98%, Thermo Scientific Chemicals
CAS: 57-71-6 | C4H7NO2 | 101.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA150375000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS3814-34-4
IUPAC Name3-(bromomethyl)pentane
Molecular FormulaC6H13Br
InChI KeyKKGUMGWNFARLSL-UHFFFAOYSA-N
SMILESCCC(CC)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.4490-1.4540 @ 20?C