(1S)-(-)-β-Pinene, 98%, Thermo Scientific Chemicals
(1S)-(-)-β-Pinene, 98%, Thermo Scientific Chemicals
(1S)-(-)-β-Pinene, 98%, Thermo Scientific Chemicals
(1S)-(-)-β-Pinene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(1S)-(-)-β-Pinene, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA150322500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4538-37-8
IUPAC Name1,4-diisocyanatobutane
Molecular FormulaC6H8N2O2
InChI KeyOVBFMUAFNIIQAL-UHFFFAOYSA-N
SMILESO=C=NCCCCN=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4510 to 1.4530 (20°C, 589 nm)
Appearance (Color)Clear light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %