4-Chlorobenzyl chloride, 99+%, Thermo Scientific Chemicals
4-Chlorobenzyl chloride, 99+%, Thermo Scientific Chemicals
4-Chlorobenzyl chloride, 99+%, Thermo Scientific Chemicals
4-Chlorobenzyl chloride, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chlorobenzyl chloride, 99+%, Thermo Scientific Chemicals
CAS: 104-83-6 | C7H6Cl2 | 161.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Catalog number FSA150242500
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS1308-96-9
IUPAC Namedieuropium(3+) trioxidandiide
Molecular FormulaEu2O3
InChI KeyAEBZCFFCDTZXHP-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Eu+3].[Eu+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light pink
Appearance (Form)Powder
Total conc. of imp.as REO=<0.1 %