Benzoic Acid, Sodium Salt, 99+%, Thermo Scientific Chemicals
Benzoic Acid, Sodium Salt, 99+%, Thermo Scientific Chemicals
Benzoic Acid, Sodium Salt, 99+%, Thermo Scientific Chemicals
Benzoic Acid, Sodium Salt, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzoic Acid, Sodium Salt, 99+%, Thermo Scientific Chemicals
CAS: 532-32-1 | C7H5NaO2 | 144.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
Packaging:
View all
Plastic Drum
Catalog number FSA148980050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 kg
Packaging:
Plastic Drum
Request bulk or custom format
Chemical Identifiers
CAS14268-66-7
IUPAC Name2H-1,3-benzodioxol-5-amine
Molecular FormulaC7H7NO2
InChI KeyXGNXYCFREOZBOL-UHFFFAOYSA-N
SMILESNC1=CC=C2OCOC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark brown to brown to black
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Melting Point (clear melt)39.0-47.0?C
FormCrystals or powder or crystalline powder or chunks or fused solid or liquid as melt