Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals
Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals
Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals
Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals
CAS: 367-51-1 | C2H3NaO2S | 114.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Packaging:
Glass bottle
Catalog number FSA148892500
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7440-06-4
IUPAC Nameplatinum
Molecular FormulaPt
InChI KeyBASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES[Pt]
View more
SpecificationsSpecification SheetSpecification Sheet
Moisture holding capacity50.00-70.00 wt.%