2-Cyanoacetamide, 99%, Thermo Scientific Chemicals
2-Cyanoacetamide, 99%, Thermo Scientific Chemicals
2-Cyanoacetamide, 99%, Thermo Scientific Chemicals
2-Cyanoacetamide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Cyanoacetamide, 99%, Thermo Scientific Chemicals
CAS: 107-91-5 | C3H4N2O | 84.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Plastic bottle
Catalog number FSA148655000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS6097-07-0
IUPAC Name2-(pent-4-yn-1-yl)-2,3-dihydro-1H-isoindole-1,3-dione
Molecular FormulaC13H11NO2
InChI KeyYNZIPXLLPFYDGM-UHFFFAOYSA-N
SMILESO=C1N(CCCC#C)C(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point87°C to 91°C
GC>=96.0 %