Nicotinic acid hydrazide, 97%, Thermo Scientific Chemicals
Nicotinic acid hydrazide, 97%, Thermo Scientific Chemicals
Nicotinic acid hydrazide, 97%, Thermo Scientific Chemicals
Nicotinic acid hydrazide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nicotinic acid hydrazide, 97%, Thermo Scientific Chemicals
CAS: 553-53-7 | C6H7N3O | 137.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Plastic bottle
Catalog number FSA148200250
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS27607-77-8
IUPAC Nametrimethylsilyl trifluoromethanesulfonate
Molecular FormulaC4H9F3O3SSi
InChI KeyFTVLMFQEYACZNP-UHFFFAOYSA-N
SMILESC[Si](C)(C)OS(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light brown
Titration with NaOH>=98.5 %
Refractive index1.3600 to 1.3620 (20°C, 589 nm)
Appearance (Form)Liquid