3-Bromothiophene, 97%, Thermo Scientific Chemicals
3-Bromothiophene, 97%, Thermo Scientific Chemicals
3-Bromothiophene, 97%, Thermo Scientific Chemicals
3-Bromothiophene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Bromothiophene, 97%, Thermo Scientific Chemicals
CAS: 872-31-1 | C4H3BrS | 163.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
Packaging:
Glass bottle
Catalog number FSA147801000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS42969-65-3
IUPAC Name(2R)-but-3-yn-2-ol
Molecular FormulaC4H6O
InChI KeyGKPOMITUDGXOSB-SCSAIBSYSA-N
SMILESC[C@@H](O)C#C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4245-1.4275 @ 20?C
FormLiquid
Optical Rotation46.4 ± 0.1? (neat)