N,N,N',N'-Tetramethyl-1,6-hexanediamine, 99%, Thermo Scientific Chemicals
N,N,N',N'-Tetramethyl-1,6-hexanediamine, 99%, Thermo Scientific Chemicals
N,N,N',N'-Tetramethyl-1,6-hexanediamine, 99%, Thermo Scientific Chemicals
N,N,N',N'-Tetramethyl-1,6-hexanediamine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N,N',N'-Tetramethyl-1,6-hexanediamine, 99%, Thermo Scientific Chemicals
CAS: 111-18-2 | C10H24N2 | 172.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
25 mL
Packaging:
Glass bottle
Catalog number FSA147492500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS452-64-2
IUPAC Name4-fluoro-1,2-dimethylbenzene
Molecular FormulaC8H9F
InChI KeyWYHBENDEZDFJNU-UHFFFAOYSA-N
SMILESCC1=CC=C(F)C=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
Refractive Index1.4795-1.4835 @ 20?C