4,4'-Sulfonyldiphenol, 99.7%
4,4'-Sulfonyldiphenol, 99.7%
4,4'-Sulfonyldiphenol, 99.7%
4,4'-Sulfonyldiphenol, 99.7%
Thermo Scientific Chemicals
4,4'-Sulfonyldiphenol, 99.7%
CAS: 80-09-1 | C12H10O4S | 250.27 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA146915000500 gPlastic bottle
Catalog number FSA146915000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS422-64-0
IUPAC Namepentafluoropropanoic acid
Molecular FormulaC3HF5O2
InChI KeyLRMSQVBRUNSOJL-UHFFFAOYSA-N
SMILESOC(=O)C(F)(F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light brown
Appearance (Form)Liquid
Infrared spectrumConforms
Titration with NaOH>=96.0 %