Chlorobenzene, 99+%, pure, Thermo Scientific Chemicals
Chlorobenzene, 99+%, pure, Thermo Scientific Chemicals
Chlorobenzene, 99+%, pure, Thermo Scientific Chemicals
Chlorobenzene, 99+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chlorobenzene, 99+%, pure, Thermo Scientific Chemicals
CAS: 108-90-7 | C6H5Cl | 112.556 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA146410025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialChlorobenzene
Melting Point-45.0°C
CAS108-90-7
Health Hazard 1Warning
Health Hazard 2GHS P Statement
Toxic to aquatic life with long lasting effects.
Harmful if inhaled.
Flammable liquid and vapor.