Triphenylmethane, 99+%, Thermo Scientific Chemicals
Triphenylmethane, 99+%, Thermo Scientific Chemicals
Triphenylmethane, 99+%, Thermo Scientific Chemicals
Triphenylmethane, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Triphenylmethane, 99+%, Thermo Scientific Chemicals
CAS: 519-73-3 | C19H16 | 244.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number FSA146381000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTriphenylmethane
Name Note99+%
CAS519-73-3
Melting Point90.0°C to 94.0°C
ColorYellow