Urea, 99.5%, for analysis, Thermo Scientific Chemicals
Urea, 99.5%, for analysis, Thermo Scientific Chemicals
Urea, 99.5%, for analysis, Thermo Scientific Chemicals
Urea, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Urea, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 57-13-6 | CH4N2O | 60.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
25 kg
Packaging:
View all
Plastic drum
Catalog number FSA140750050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS10213-10-2
IUPAC Namedisodium dioxotungstenbis(olate) dihydrate
Molecular FormulaH4Na2O6W
InChI KeyWPZFLQRLSGVIAA-UHFFFAOYSA-N
SMILESO.O.[Na+].[Na+].[O-][W]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)94.1% min
For analysis
RUO – Research Use Only