Tyramine, 97%, Thermo Scientific Chemicals
Tyramine, 97%, Thermo Scientific Chemicals
Tyramine, 97%, Thermo Scientific Chemicals
Tyramine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tyramine, 97%, Thermo Scientific Chemicals
CAS: 51-67-2 | C8H11NO | 137.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA140610050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS75-09-2
IUPAC Namedichloromethane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-UHFFFAOYSA-N
SMILESClCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=99.8 %
Water=<0.005 % (Coulometric)
StabilizerApprox. 50 ppm Amylene