Triphenyl phosphite, 99%, Thermo Scientific Chemicals
Triphenyl phosphite, 99%, Thermo Scientific Chemicals
Triphenyl phosphite, 99%, Thermo Scientific Chemicals
Triphenyl phosphite, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Triphenyl phosphite, 99%, Thermo Scientific Chemicals
CAS: 101-02-0 | C18H15O3P | 310.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
1 L
2.5 L
Packaging:
View all
Plastic drum
Catalog number FSA140440100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 L
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS290-87-9
IUPAC Name1,3,5-triazine
Molecular FormulaC3H3N3
InChI KeyJIHQDMXYYFUGFV-UHFFFAOYSA-N
SMILESC1=NC=NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤1.5%
Melting Point (clear melt)72.0-83.0?C
Assay (GC)≥96.0%