2,4,4-Trimethyl-1-pentene, 99%, Thermo Scientific Chemicals
2,4,4-Trimethyl-1-pentene, 99%, Thermo Scientific Chemicals
2,4,4-Trimethyl-1-pentene, 99%, Thermo Scientific Chemicals
2,4,4-Trimethyl-1-pentene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,4,4-Trimethyl-1-pentene, 99%, Thermo Scientific Chemicals
CAS: 107-39-1 | C8H16 | 112.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number FSA140181000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7365-45-9
IUPAC Name2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
Molecular FormulaC8H18N2O4S
InChI KeyJKMHFZQWWAIEOD-UHFFFAOYSA-N
SMILESOCCN1CCN(CCS(O)(=O)=O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
UVat 260 nm A: =<0.08
UVat 280 nm A: =<0.05
Solubility(5 % in water) Clear colorless
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals