1,2,4-Trimethylbenzene, 98%, Thermo Scientific Chemicals
1,2,4-Trimethylbenzene, 98%, Thermo Scientific Chemicals
1,2,4-Trimethylbenzene, 98%, Thermo Scientific Chemicals
1,2,4-Trimethylbenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,2,4-Trimethylbenzene, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number FSA140090010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS60-18-4
IUPAC Name(2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid
Molecular FormulaC9H11NO3
InChI KeyOUYCCCASQSFEME-SVGMAFHSNA-N
SMILESN[C@@H](CC1=CC=C(O)C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=99.0 % (on dried substance)
Loss on drying=<0.5 % (1 g, 105°C)
Appearance (Form)Powder
Infrared spectrumConforms
Heavy metals (as Pb)=<10 ppm