4',5,7-Trihydroxyflavanone, 97%
4',5,7-Trihydroxyflavanone, 97%
4',5,7-Trihydroxyflavanone, 97%
4',5,7-Trihydroxyflavanone, 97%
Thermo Scientific Chemicals
4',5,7-Trihydroxyflavanone, 97%
CAS: 480-41-1 | C15H12O5 | 272.26 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA13982010010 gGlass bottle
Catalog number FSA139820100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4', 5, 7-Trihydroxyflavanone
Name Note97%
CAS480-41-1
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning