o-Toluidine, 99%, Thermo Scientific Chemicals
o-Toluidine, 99%, Thermo Scientific Chemicals
o-Toluidine, 99%, Thermo Scientific Chemicals
o-Toluidine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
o-Toluidine, 99%, Thermo Scientific Chemicals
CAS: 95-53-4 | C7H9N | 107.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA139085000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1124-63-6
IUPAC Name3-cyclohexylpropan-1-ol
Molecular FormulaC9H18O
InChI KeyCLYAQFSQLQTVNO-UHFFFAOYSA-N
SMILESOCCCC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.4650 to 1.4670 (20°C, 589 nm)
Appearance (Form)Viscous liquid
GC>=98.5 %