p-Toluenesulfonyl chloride, 99+%, Thermo Scientific Chemicals
p-Toluenesulfonyl chloride, 99+%, Thermo Scientific Chemicals
p-Toluenesulfonyl chloride, 99+%, Thermo Scientific Chemicals
p-Toluenesulfonyl chloride, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Toluenesulfonyl chloride, 99+%, Thermo Scientific Chemicals
CAS: 98-59-9 | C7H7ClO2S | 190.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
2 kg
Packaging:
Glass bottle
Catalog number FSA139031000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS20445-31-2
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid or clear liquid or viscous liquid as melt
Optical Rotation71.0 ± 2.0? (C=2 in methanol)
Appearance (Color)Colorless to white
Identification (FTIR)Conforms
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%