Thiophene, 99+%, extra pure, Thermo Scientific Chemicals
Thiophene, 99+%, extra pure, Thermo Scientific Chemicals
Thiophene, 99+%, extra pure, Thermo Scientific Chemicals
Thiophene, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thiophene, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 110-02-1 | C4H4S | 84.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Glass bottle
Catalog number FSA138825000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS50528-80-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥80.0%
Refractive Index1.4715-1.4795 @ 20?C