Sulfuric Acid, 96% Solution in Water, Extra Pure, Thermo Scientific Chemicals
Sulfuric Acid, 96% Solution in Water, Extra Pure, Thermo Scientific Chemicals
Sulfuric Acid, 96% Solution in Water, Extra Pure, Thermo Scientific Chemicals
Sulfuric Acid, 96% Solution in Water, Extra Pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sulfuric Acid, 96% Solution in Water, Extra Pure, Thermo Scientific Chemicals
CAS: 7664-93-9 | H2O4S | 98.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Plastic Bottle
Catalog number FSA133610026
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS5122-94-1
IUPAC Name{[1,1'-biphenyl]-4-yl}boronic acid
Molecular FormulaC12H11BO2
InChI KeyXPEIJWZLPWNNOK-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(C=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Powder or crystalline powder
HPLC>=96.0 %