D-Sorbitol, 97%
D-Sorbitol, 97%
D-Sorbitol, 97%
Thermo Scientific Chemicals
D-Sorbitol, 97%
CAS: 50-70-4 | C6H14O6 | 182.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
1 kg
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA132735000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS10025-77-1
IUPAC Nameiron(3+) hexahydrate trichloride
Molecular FormulaCl3FeH12O6
InChI KeyNQXWGWZJXJUMQB-UHFFFAOYSA-K
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange-brown
Nitrate (NO3)=<100 ppm
Sulfate (SO4)=<100 ppm
Appearance (Form)Adhering crystalline powder and/or lumps
Phosphorous compounds (as PO4)=<100 ppm