Piperonyl alcohol, 98%, Thermo Scientific Chemicals
Piperonyl alcohol, 98%, Thermo Scientific Chemicals
Piperonyl alcohol, 98%, Thermo Scientific Chemicals
Piperonyl alcohol, 98%, Thermo Scientific Chemicals
Piperonyl alcohol, 98%, Thermo Scientific Chemicals
CAS: 495-76-1 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA131350250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS591-78-6
IUPAC Namehexan-2-one
Molecular FormulaC6H12O
InChI KeyQQZOPKMRPOGIEB-UHFFFAOYSA-N
SMILESCCCCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=97.5 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.3995 to 1.4015 (20°C, 589 nm)