Phenyl sulfone, 97%, Thermo Scientific Chemicals
Phenyl sulfone, 97%, Thermo Scientific Chemicals
Phenyl sulfone, 97%, Thermo Scientific Chemicals
Phenyl sulfone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenyl sulfone, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Plastic bottle
Catalog number FSA130962500
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7440-57-5
IUPAC Namegold
Molecular FormulaAu
InChI KeyPCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES[Au]
View more
SpecificationsSpecification SheetSpecification Sheet
FormWire
Diameter0.45-0.55mm
Total Metal Impurities≤ 0.0015%
CommentGold