4-Phenoxyaniline, 97%, Thermo Scientific Chemicals
4-Phenoxyaniline, 97%, Thermo Scientific Chemicals
4-Phenoxyaniline, 97%, Thermo Scientific Chemicals
4-Phenoxyaniline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Phenoxyaniline, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA130231000
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS104-52-9
IUPAC Name(3-chloropropyl)benzene
Molecular FormulaC9H11Cl
InChI KeyXZBXAYCCBFTQHH-UHFFFAOYSA-N
SMILESClCCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to brown
Assay (GC)≥96.0%
FormLiquid
Refractive Index1.5205-1.5245 @ 20°C