Orcinol Monohydrate, 99%, Thermo Scientific Chemicals
Orcinol Monohydrate, 99%, Thermo Scientific Chemicals
Orcinol Monohydrate, 99%, Thermo Scientific Chemicals
Orcinol Monohydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Orcinol Monohydrate, 99%, Thermo Scientific Chemicals
CAS: 6153-39-5 | C7H8O2·H2O | 142.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA129550250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS94-36-0
IUPAC Namebenzoyl benzenecarboperoxoate
Molecular FormulaC14H10O4
InChI KeyOMPJBNCRMGITSC-UHFFFAOYSA-N
SMILESO=C(OOC(=O)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Moist granular powder
Appearance (Color)White
Impurity=<0.25 % Inorganic and organic hydrolizable chloride
Titration Iodometric73 to 77 %