n-Octylamine, 99+%, Thermo Scientific Chemicals
n-Octylamine, 99+%, Thermo Scientific Chemicals
n-Octylamine, 99+%, Thermo Scientific Chemicals
n-Octylamine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Octylamine, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
Glass bottle
Catalog number FSA129495000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS12230-71-6
IUPAC Namebarium(2+) octahydrate dihydroxide
Molecular FormulaBaH18O10
InChI KeyZUDYPQRUOYEARG-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals or flakes
Titration with HCl>=98.0 %
Heavy metals (as Pb)=<5 ppm
Insoluble matter=<0.01 % (in HCl)
Barium carbonate=<2 %