2-Nitroaniline, 98%, Thermo Scientific Chemicals
2-Nitroaniline, 98%, Thermo Scientific Chemicals
2-Nitroaniline, 98%, Thermo Scientific Chemicals
2-Nitroaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Nitroaniline, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA128351000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4830-93-7
IUPAC Name(4-chlorobutyl)benzene
Molecular FormulaC10H13Cl
InChI KeyFLLZCZIHURYEQP-UHFFFAOYSA-N
SMILESClCCCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5160-1.5210 @ 20?C
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
Indicator
RUO – Research Use Only